Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5730508B09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027482.3 | NP_081758.1 | 1, 2 | 285 | ||
GenBank mRNA | AK012470.1 | - | 1, 2 | 256 | ||
AK017758.1 | - | 1, 2 | 248 | |||
AK162420.1 | - | 1, 2 | 262 | |||
AK163003.1 | - | 1, 2 | 252 | |||
BC039568.1 | - | 1, 2 | 12 | |||
BC116791.1 | - | 1, 2 | 232 | 40126110 | ||
BC116793.1 | - | 1, 2 | 232 | 40126112 |