Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027490.1 | NP_081766.1 | 4, 5 | 565 | ||
GenBank mRNA | AK017809.1 | - | 4, 5 | 557 | ||
AK152160.1 | - | 4, 5 | 538 | |||
AK153105.1 | - | 4, 5 | 567 | |||
AK153264.1 | - | 4, 5 | 538 | |||
AK153310.1 | - | 4, 5 | 538 | |||
AK166560.1 | - | 4, 5 | 544 |