Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Utp20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175158.3 | NP_780367.2 | 59, 60 | 8107 | ||
GenBank mRNA | AB052760.1 | - | 17, 18 | 2308 | ||
AK014430.1 | - | 5, 6 | 735 | |||
AK047081.1 | - | 2158 | ||||
BC048955.1 | - | 29, 30 | 3972 |