Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027513.1 | NP_081789.1 | 11 | 1529 | ||
GenBank mRNA | AK014419.1 | - | 11 | 1529 | ||
AK129093.1 | - | 11 | 1714 | |||
BC058729.1 | - | 10 | 1498 | |||
BC060139.1 | - | 8 | 1042 | |||
BC060711.1 | - | 8 | 1042 | |||
BC141333.1 | - | 11 | 1715 | 9055945 |