Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nup205 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027513.1 | NP_081789.1 | 40 | 5594 | ||
GenBank mRNA | AK129093.1 | - | 40 | 5779 | ||
BC027832.1 | - | 4 | 644 | |||
BC037401.1 | - | 26 | 3351 | |||
BC045524.1 | - | 8 | 837 | |||
BC058729.1 | - | 39 | 5563 | |||
BC060139.1 | - | 37 | 5107 | |||
BC060223.1 | - | 27 | 3876 | |||
BC060531.1 | - | 11 | 1273 | |||
BC060711.1 | - | 37 | 5107 | |||
BC141333.1 | - | 40 | 5780 | 9055945 |