Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dclk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195496.1 | NP_001182425.1 | 16 | 2751 | ||
NM_001195497.1 | NP_001182426.1 | 15 | 2703 | |||
NM_001195498.1 | NP_001182427.1 | 16 | 2754 | |||
NM_001195499.1 | NP_001182428.1 | 16 | 2751 | |||
NM_027539.5 | NP_081815.3 | 15 | 2706 | |||
GenBank mRNA | AB198721.1 | - | 16 | 2108 | ||
AK049179.1 | - | 15 | 2155 | |||
AK082633.1 | - | 14 | 2121 | |||
AK188709.1 | - | 2 | 141 | |||
AY968049.1 | - | 16 | 2039 | |||
AY968050.1 | - | 16 | 2042 | |||
AY968051.1 | - | 16 | 2039 | |||
BC052071.1 | - | 8 | 747 | |||
BC056921.1 | - | 15 | 2674 | 6822681 |