Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nolc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039351.2 | NP_001034440.1 | 11 | 1983 | ||
NM_001039352.2 | NP_001034441.1 | 11 | 1989 | |||
NM_001039353.2 | NP_001034442.1 | 11 | 1986 | |||
NM_053086.3 | NP_444316.2 | 11 | 1986 | |||
GenBank mRNA | AK014336.1 | - | 11 | 1888 | ||
AK029150.1 | - | 11 | 1834 | |||
AK034817.1 | - | 11 | 1863 | |||
AK129040.1 | - | 11 | 1810 | |||
AK147118.1 | - | 11 | 1859 | |||
AK150332.1 | - | 11 | 1820 | |||
AK151031.1 | - | 11 | 1820 | |||
AK152941.1 | - | 11 | 1820 | |||
AK166753.1 | - | 11 | 1892 |