Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nolc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039351.2 | NP_001034440.1 | 12 | 2086 | ||
NM_001039352.2 | NP_001034441.1 | 12 | 2092 | |||
NM_001039353.2 | NP_001034442.1 | 12 | 2089 | |||
NM_053086.3 | NP_444316.2 | 12 | 2089 | |||
GenBank mRNA | AK014336.1 | - | 12 | 1991 | ||
AK029150.1 | - | 12 | 1937 | |||
AK034817.1 | - | 12 | 1966 | |||
AK129040.1 | - | 12 | 1913 | |||
AK147118.1 | - | 12 | 1962 | |||
AK150332.1 | - | 12 | 1923 | |||
AK151031.1 | - | 12 | 1923 | |||
AK152941.1 | - | 12 | 1923 | |||
AK166753.1 | - | 12 | 1995 |