Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arrdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027560.1 | NP_081836.1 | 4 | 840 | ||
GenBank mRNA | AK014582.1 | - | 4 | 840 | ||
AK046754.1 | - | 4 | 550 | |||
AK135740.1 | - | 4 | 656 | |||
AK137175.1 | - | 4 | 657 | |||
AK144323.1 | - | 5 | 574 | |||
BC036384.1 | - | 4 | 638 | 1067080 | ||
BC043313.1 | - | 1 | 194 | |||
BC043469.1 | - | 1 | 194 | |||
BC054826.1 | - | 1 | 200 | |||
BC094060.1 | - | 3 | 720 | |||
DQ861995.1 | - | 4 | 504 |