Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025428.1 | NP_001020599.1 | 2 | 427 | ||
NM_001025429.1 | NP_001020600.1 | 2 | 427 | |||
NM_001025430.1 | NP_001020601.1 | 2 | 427 | |||
NM_001199203.1 | NP_001186132.1 | 5 | 876 | |||
NM_001199204.1 | NP_001186133.1 | 5 | 876 | |||
NM_001199205.1 | NP_001186134.1 | 2 | 427 | |||
NM_027569.2 | NP_081845.2 | 5 | 876 | |||
GenBank mRNA | AB047782.1 | - | 5 | 796 | ||
AF327451.1 | - | 5 | 858 | |||
AK030165.1 | - | 5 | 697 | |||
AK147431.1 | - | 2 | 484 | |||
AK147537.1 | - | 2 | 503 | |||
AK161623.1 | - | 4 | 756 | |||
AK172961.1 | - | 2 | 409 | |||
AY823270.1 | - | 5 | 420 | |||
BC052058.1 | - | 2 | 364 | |||
BC060100.1 | - | 2 | 361 | 6417257 | ||
BC094670.1 | - | 2 | 355 | 30618738 | ||
CK781075.1 | - | 2 | 391 | |||
FJ042496.1 | - | 5 | 753 |