Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc63 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027581.1 | NP_081857.1 | 9 | 1770 | ||
GenBank mRNA | AK014850.1 | - | 8 | 1502 | ||
BC145796.1 | - | 9 | 1765 | 40131099 |