Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atoh8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153778.3 | NP_722473.1 | 1 | 990 | ||
GenBank mRNA | AB046527.1 | - | 1 | 992 | ||
AB046528.1 | - | 1 | 688 | |||
AB049066.1 | - | 1 | 577 | |||
AY349615.1 | - | 1 | 698 | |||
BC023684.1 | - | 1 | 658 | 5343472 |