Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027708.1 | NP_081984.1 | 9 | 1408 | ||
GenBank mRNA | AK016870.1 | - | 9 | 1409 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AK134703.1 | - | 307 | Chr. - - [-] on Build |