Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spata24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027733.5 | NP_082009.3 | 2 | 209 | ||
NM_029485.2 | NP_083761.1 | 2 | 209 | |||
GenBank mRNA | AK016341.1 | - | 2 | 215 | ||
AY730532.1 | - | 2 | 209 | |||
BC060950.1 | - | 2 | 162 | 6773733 | ||
BU961109.1 | - | 2 | 212 |