Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf148 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027754.1 | NP_082030.1 | 1 | 751 | ||
GenBank mRNA | AK017027.1 | - | 1 | 752 | ||
AY155440.1 | - | 1 | 703 |