Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 5530601H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017459.1 | - | 302 | |||
AK043766.1 | - | 338 | ||||
AK047017.1 | - | 338 | ||||
AK080296.1 | - | 3, 4 | 333 | |||
AK084653.1 | - | 3, 4 | 338 | |||
AK084980.1 | - | 3, 4 | 317 | |||
AK085098.1 | - | 3, 4 | 310 | |||
AK085485.1 | - | 3, 4 | 304 | |||
AK144934.1 | - | 3, 4 | 239 | |||
AK153718.1 | - | 177 | ||||
AK157237.1 | - | 296 | ||||
AK164650.1 | - | 3, 4 | 366 | |||
BC019699.1 | - | 180 | ||||
BC049927.1 | - | 154 | ||||
BC055463.1 | - | 3, 4 | 212 |