Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyp2u1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027816.3 | NP_082092.2 | 1 | 302 | ||
GenBank mRNA | AK018458.1 | - | 1 | 278 | ||
AK041477.1 | - | 1 | 303 | |||
AK142740.1 | - | 1 | 242 |
Gene Symbol: LOC100503594 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090212.1 | - | 281 |