Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027823.1 | NP_082099.1 | 5 | 658 | ||
GenBank mRNA | AK082857.1 | - | 5 | 747 | ||
BC137891.1 | - | 5 | 658 | 8860910 | ||
BC137892.1 | - | 5 | 658 | 8860911 |