Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 4930506M07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114312.1 | NP_001107784.1 | 8 | 1202 | ||
NM_175172.4 | NP_780381.1 | 8 | 1202 | |||
GenBank mRNA | AK015721.1 | - | 1 | 134 | ||
AK039224.1 | - | 4 | 273 | |||
AK082304.1 | - | 8 | 1203 | |||
AK220405.1 | - | 8 | 1232 | |||
BC030338.1 | - | 8 | 774 | 2599101 | ||
EF055486.1 | - | 8 | 645 |