Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rarres2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027852.2 | NP_082128.1 | 2 | 236 | ||
GenBank mRNA | AK002298.1 | - | 2 | 237 | ||
BC015275.1 | - | 2 | 212 | |||
BC038914.1 | - | 2 | 256 | 5100583 |