Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ubiad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027873.2 | NP_082149.1 | 1 | 545 | ||
GenBank mRNA | AK004550.1 | - | 1 | 545 | ||
AK034787.1 | - | 1 | 572 | |||
AK078442.2 | - | 1 | 546 | |||
BC015303.1 | - | 1 | 253 | 4241204 | ||
BC071203.1 | - | 1 | 534 | 3667534 |