Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpn2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027904.3 | NP_082180.2 | 2 | 874 | ||
GenBank mRNA | AK005049.1 | - | 2 | 876 | ||
BC025836.1 | - | 2 | 841 | |||
BC081550.1 | - | 1 | 393 | |||
BC138287.1 | - | 2 | 961 | 8861306 | ||
BC138288.1 | - | 2 | 961 | 8861307 |