Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023625.4 | NP_076114.2 | 9, 10 | 1335 | ||
GenBank mRNA | AF217003.1 | - | 9, 10 | 1316 | ||
AK004973.1 | - | 9, 10 | 1338 | |||
AK030234.1 | - | 9, 10 | 1287 | |||
AK032930.1 | - | 9, 10 | 1331 | |||
AK042089.1 | - | 1331 | ||||
AK043608.1 | - | 9, 10 | 1325 | |||
AK150201.1 | - | 9, 10 | 1324 | |||
AK164220.1 | - | 9, 10 | 1316 | |||
AK170387.1 | - | 9, 10 | 1327 | |||
AK170631.1 | - | 9, 10 | 1315 | |||
AK188200.1 | - | 1, 2 | 75 | |||
AK209064.1 | - | 1, 2 | 75 | |||
BC026395.1 | - | 9, 10 | 1313 | |||
BC038605.1 | - | 9, 10 | 1316 | 4921245 | ||
BC048826.1 | - | 9, 10 | 1322 | 4196604 |