Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Shroom1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290789.1 | NP_001277718.1 | 4 | 1338 | ||
NM_027917.3 | NP_082193.2 | 5 | 1527 | |||
GenBank mRNA | AK004934.1 | - | 5 | 1454 | ||
AK035462.1 | - | 3 | 2464 | |||
AK085693.1 | - | 4 | 1299 | |||
AK134332.1 | - | 5 | 1455 | |||
BC005761.1 | - | 4 | 1313 | 3601742 | ||
BC031598.1 | - | 4 | 1230 | 3497170 |