Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ing3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023626.4 | NP_076115.3 | 1, 2 | 170 | ||
GenBank mRNA | AK080787.1 | - | 1, 2 | 139 | ||
AK131993.1 | - | 1, 2 | 133 | |||
AY007791.1 | - | 1, 2 | 139 | |||
BC018342.1 | - | 1, 2 | 120 | 4010253 |