Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: March8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK005032.2 | - | 7 | 836 | ||
AK154976.1 | - | 8 | 1028 | |||
AK165647.1 | - | 7 | 828 | |||
AK208723.1 | - | 2 | 178 | |||
BC050908.1 | - | 8 | 1011 | 6416955 | ||
BC053090.1 | - | 7 | 1096 | 5709246 |