Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zswim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027964.2 | NP_082240.1 | 9 | 1445 | ||
GenBank mRNA | AK006367.1 | - | 9 | 1446 | ||
BC120837.1 | - | 9 | 1425 | 40129760 | ||
BC144860.1 | - | 9 | 1413 | |||
EU233996.1 | - | 7 | 1560 |