Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdadc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168535.1 | NP_001162007.1 | 8 | 1520 | ||
NM_001168536.1 | NP_001162008.1 | 8 | 1520 | |||
NM_001168537.1 | NP_001162009.1 | 8 | 1520 | |||
NM_001168538.1 | NP_001162010.1 | 8 | 1520 | |||
NM_027986.3 | NP_082262.1 | 8 | 1520 | |||
GenBank mRNA | AK009280.1 | - | 8 | 1512 | ||
AK032600.1 | - | 8 | 1499 | |||
AK032813.1 | - | 8 | 1506 | |||
AK038926.1 | - | 8 | 1500 | |||
AK039148.1 | - | 8 | 1508 | |||
BC004588.1 | - | 4 | 535 | |||
BC006901.1 | - | 8 | 1482 | 3600168 | ||
BE648840.1 | - | 2 | 267 |