Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Paqr7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285845.1 | NP_001272774.1 | 3 | 516 | ||
NM_001285846.1 | NP_001272775.1 | 4 | 442 | |||
NM_001285847.1 | NP_001272776.1 | 3 | 405 | |||
NM_001285849.1 | NP_001272778.1 | 3 | 289 | |||
NM_027995.3 | NP_082271.1 | 2 | 321 | |||
GenBank mRNA | AF313618.1 | - | 2 | 75 | ||
AK009450.1 | - | 2 | 106 | |||
AK078685.1 | - | 2 | 79 | |||
AK161542.1 | - | 3 | 163 | |||
AK170307.1 | - | 4 | 226 | |||
AY278956.1 | - | 1 | 14 | |||
AY424296.1 | - | 1 | 50 | |||
BC022922.1 | - | 1 | 50 | 4239768 | ||
BY210355.1 | - | 2 | 323 | |||
BY288540.1 | - | 3 | 290 |
Set Membership: |
G-protein coupled receptor |