Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem123 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133739.2 | NP_598500.1 | 3 | 338 | ||
GenBank mRNA | AK150361.1 | - | 3 | 286 | ||
AK150365.1 | - | 4 | 366 | |||
AK151044.1 | - | 3 | 306 | |||
AK152180.1 | - | 3 | 286 | |||
AK152423.1 | - | 3 | 251 | |||
AK153122.1 | - | 3 | 251 | |||
AK153378.1 | - | 4 | 366 | |||
AK153539.1 | - | 3 | 286 | |||
AK155767.1 | - | 3 | 306 | |||
AK157001.1 | - | 4 | 263 | |||
AK162610.1 | - | 3 | 334 | |||
AK165671.1 | - | 3 | 305 | |||
AK170823.1 | - | 3 | 297 | |||
AK172352.1 | - | 3 | 267 | |||
AK172553.1 | - | 3 | 267 | |||
AK172701.1 | - | 3 | 267 | |||
BC010292.1 | - | 3 | 287 | 3153174 | ||
BC086319.1 | - | 3 | 295 | 5293409 |