Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nkiras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028024.2 | NP_082300.1 | 4 | 494 | ||
GenBank mRNA | AK010371.1 | - | 4 | 494 | ||
AK015918.1 | - | 499 | ||||
AK052645.1 | - | 4 | 494 | |||
AK083475.1 | - | 4 | 537 | |||
AK149696.1 | - | 4 | 638 | |||
AK169878.1 | - | 4 | 519 | |||
BC013469.1 | - | 3 | 447 | 4219892 | ||
BC024398.1 | - | 3 | 437 | 5003001 |