Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snx10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127348.1 | NP_001120820.1 | 5 | 446 | ||
NM_001127349.1 | NP_001120821.1 | 5 | 451 | |||
NM_028035.4 | NP_082311.3 | 5 | 549 | |||
GenBank mRNA | AK010399.1 | - | 5 | 394 | ||
AK028163.1 | - | 5 | 550 | |||
AK150704.1 | - | 5 | 454 | |||
AK150878.1 | - | 5 | 368 | |||
AK150999.1 | - | 5 | 454 | |||
AK151320.1 | - | 5 | 454 | |||
AK151520.1 | - | 5 | 454 | |||
AK152004.1 | - | 5 | 372 | |||
AK152825.1 | - | 5 | 426 | |||
AK152949.1 | - | 5 | 442 | |||
AK153127.1 | - | 5 | 454 | |||
AK153356.1 | - | 5 | 404 | |||
AK166121.1 | - | 5 | 419 | |||
AK169110.1 | - | 5 | 385 | |||
BC010334.1 | - | 5 | 383 | 2655824 | ||
CT010276.1 | - | 4 | 296 |