Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028048.2 | NP_082324.1 | 5 | 1341 | ||
GenBank mRNA | AK007453.1 | - | 5 | 1343 | ||
AK035739.1 | - | 1312 | ||||
AK171131.1 | - | 5 | 1348 | |||
BC004569.1 | - | 5 | 781 | |||
BC019996.1 | - | 5 | 1283 | 4207271 |