Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001141975.1 | NP_001135447.1 | 11, 12 | 1478 | ||
NM_001141976.1 | NP_001135448.1 | 11, 12 | 1494 | |||
NM_001141977.1 | NP_001135449.1 | 11, 12 | 1682 | |||
NM_001141978.1 | NP_001135450.1 | 11, 12 | 1481 | |||
NM_028109.4 | NP_082385.3 | 11, 12 | 1669 | |||
GenBank mRNA | AK011311.2 | - | 8, 9 | 1037 | ||
AK090101.1 | - | 11, 12 | 1432 | |||
AK153957.1 | - | 11, 12 | 1416 | |||
AK167030.1 | - | 8, 9 | 1118 | |||
BC060619.1 | - | 11, 12 | 1570 | 6812536 |