Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thumpd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028138.1 | NP_082414.1 | 9 | 1126 | ||
GenBank mRNA | AK012806.1 | - | 9 | 1109 |