Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 1600014C10Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085385.1 | NP_001078854.1 | 2 | 404 | ||
NM_028166.3 | NP_082442.1 | 2 | 212 | |||
GenBank mRNA | AK005449.1 | - | 2 | 184 | ||
AK032135.1 | - | 2 | 182 | |||
AK032342.1 | - | 2 | 183 | |||
AK146474.1 | - | 2 | 192 | |||
BC010476.1 | - | 2 | 168 | 3673971 | ||
BC019834.1 | - | 2 | 335 | 5123719 | ||
BC085480.1 | - | 2 | 150 | 6401538 | ||
BC092099.1 | - | 571 | 5136977 | |||
CK335181.1 | - | 2 | 164 |