Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccpg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114328.2 | NP_001107800.1 | 2, 3 | 237 | ||
NM_001286544.1 | NP_001273473.1 | 2, 3 | 237 | |||
NM_001286545.1 | NP_001273474.1 | 2, 3 | 237 | |||
NM_001286546.1 | NP_001273475.1 | 2, 3 | 237 | |||
NM_028181.5 | NP_082457.3 | 2, 3 | 237 | |||
GenBank mRNA | AK007973.1 | - | 2, 3 | 134 | ||
AK032645.1 | - | 2, 3 | 123 | |||
AK076325.2 | - | 2, 3 | 164 | |||
AK128894.1 | - | 2, 3 | 163 | |||
AK134675.1 | - | 2, 3 | 134 | |||
BC043049.1 | - | 2, 3 | 136 | 5704185 | ||
BC082602.1 | - | 2, 3 | 138 | 30663407 | ||
CN721780.1 | - | 3, 4 | 350 |