Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Plekhf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024413.2 | NP_077724.2 | 2 | 110 | ||
GenBank mRNA | AK143976.1 | - | 99 | |||
AK157990.1 | - | 2 | 99 | |||
AK170283.1 | - | 2 | 109 | |||
AK171398.1 | - | 2 | 99 | |||
AW227287.1 | - | 2 | 110 | |||
BC002120.1 | - | 1 | 75 | 3491671 |