Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cyth4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028195.3 | NP_082471.2 | 4 | 339 | ||
GenBank mRNA | AF079972.1 | - | 4 | 270 | ||
AK030956.1 | - | 4 | 1224 | |||
AK036679.1 | - | 4 | 245 | |||
AK152306.1 | - | 4 | 245 | |||
AK152909.1 | - | 4 | 245 | |||
AK157026.1 | - | 4 | 245 | |||
AK220198.1 | - | 4 | 268 | |||
BC018505.1 | - | 3 | 163 | |||
BC139054.1 | - | 4 | 312 | 8862076 | ||
BC139056.1 | - | 4 | 312 | 8862078 | ||
BC145528.1 | - | 4 | 312 | |||
BY161747.1 | - | 4 | 343 |