Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdkn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028222.1 | NP_082498.1 | 2 | 130 | ||
GenBank mRNA | AK010426.1 | - | 2 | 88 | ||
AK033341.1 | - | 2 | 94 | |||
AK146312.1 | - | 2 | 46 | |||
BC049694.1 | - | 2 | 114 | 6705916 |