Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Faim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038658.2 | NP_001033747.1 | 4 | 896 | ||
NM_028224.4 | NP_082500.2 | 4 | 932 | |||
GenBank mRNA | AF468028.1 | - | 3 | 361 | ||
AK013476.1 | - | 4 | 335 | |||
AK030513.1 | - | 4 | 419 | |||
AK080839.1 | - | 4 | 445 | |||
AK081252.1 | - | 4 | 450 | |||
AK129246.1 | - | 4 | 445 | |||
AK136791.1 | - | 4 | 414 | |||
AK147343.1 | - | 4 | 450 | |||
AK158960.1 | - | 4 | 725 | |||
BC072560.1 | - | 4 | 932 |