Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rbm12b1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028226.2 | NP_082502.2 | 3 | 2858 | ||
GenBank mRNA | AK013872.1 | - | 1 | 1953 | ||
AK030861.1 | - | 4 | 2566 | |||
BC031713.1 | - | 1 | 2455 | |||
BC050844.1 | - | 3 | 2659 | 6513134 | ||
BC054081.1 | - | 1 | 1591 | |||
BP767112.1 | - | 1 | 478 |