Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc71 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133744.4 | NP_598505.1 | 2 | 603 | ||
GenBank mRNA | AK011229.1 | - | 2 | 569 | ||
AK044601.1 | - | 2 | 569 | |||
AK079333.1 | - | 2 | 576 | |||
AK085544.1 | - | 2 | 595 | |||
AK086560.1 | - | 2 | 569 | |||
AK136982.1 | - | 2 | 556 | |||
AK149878.1 | - | 2 | 569 | |||
AK161396.1 | - | 2 | 679 | |||
BC018518.1 | - | 2 | 554 | 2647509 |