Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem87b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028248.2 | NP_082524.2 | 6 | 575 | ||
GenBank mRNA | AK011959.1 | - | 6 | 496 | ||
AK050203.1 | - | 6 | 567 | |||
AK164504.1 | - | 6 | 557 | |||
AK171811.1 | - | 7 | 491 | |||
AK172611.1 | - | 7 | 491 | |||
BC031488.1 | - | 6 | 531 |