Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013024.2 | NP_001013042.1 | 18 | 2225 | ||
GenBank mRNA | AK135141.1 | - | 18 | 2277 | ||
BC090999.1 | - | 18 | 2225 | 6754549 |