Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 2700029M09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028299.1 | NP_082575.1 | 3 | 276 | ||
GenBank mRNA | AK012304.1 | - | 3 | 252 | ||
AK150453.1 | - | 3 | 262 | |||
AK157260.1 | - | 3 | 262 | |||
BC042740.1 | - | 3 | 276 | 5024951 |