Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146024.1 | NP_001139496.1 | 5 | 1042 | ||
NM_028316.3 | NP_082592.2 | 4 | 922 | |||
GenBank mRNA | AK018079.1 | - | 5 | 1040 | ||
AK089886.1 | - | 4 | 891 | |||
AK169973.1 | - | 4 | 934 | |||
AK220307.1 | - | 360 | ||||
BC022736.1 | - | 1 | 260 | |||
BC023396.2 | - | 5 | 1108 | |||
BC037180.1 | - | 4 | 890 | |||
BC139359.1 | - | 4 | 878 | 8862381 | ||
BC139361.1 | - | 4 | 878 | 8862383 |