Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp444 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146024.1 | NP_001139496.1 | 5 | 1145 | ||
NM_028316.3 | NP_082592.2 | 4 | 1025 | |||
GenBank mRNA | AK018079.1 | - | 5 | 1143 | ||
AK089886.1 | - | 4 | 994 | |||
AK169973.1 | - | 4 | 1037 | |||
AK220307.1 | - | 463 | ||||
BC022736.1 | - | 1 | 363 | |||
BC139359.1 | - | 4 | 981 | 8862381 | ||
BC139361.1 | - | 4 | 981 | 8862383 |