Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1qtnf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001204152.1 | NP_001191081.1 | 2 | 359 | ||
NM_001204153.1 | NP_001191082.1 | 2 | 359 | |||
NM_028331.5 | NP_082607.3 | 2 | 359 | |||
GenBank mRNA | AK012868.1 | - | 2 | 301 | ||
AK051058.1 | - | 2 | 273 | |||
AK154407.1 | - | 2 | 336 | |||
AK198051.1 | - | 1 | 84 | |||
BC025489.1 | - | 2 | 302 | |||
BC071187.1 | - | 2 | 326 | 6395221 | ||
DQ002399.1 | - | 2 | 325 |