Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crtac1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145123.4 | NP_660105.3 | 7, 8 | 1157 | ||
GenBank mRNA | AJ421516.1 | - | 7, 8 | 1007 | ||
AK032328.1 | - | 7, 8 | 1022 | |||
AK049801.1 | - | 7, 8 | 1128 | |||
AK083221.1 | - | 7, 8 | 1161 | |||
AK149020.1 | - | 7, 8 | 1163 | |||
AK149069.1 | - | 7, 8 | 1158 | |||
BC024472.1 | - | 4, 5 | 593 | 4976509 |